Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109913.1 | NP_001103383.1 | 2 | 208 | ||
NM_029804.3 | NP_084080.1 | 2 | 208 | |||
GenBank mRNA | AK011521.1 | - | 2 | 154 | ||
AK051976.1 | - | 2 | 154 | |||
AK133911.1 | - | 2 | 143 | |||
AK145343.1 | - | 2 | 146 | |||
AK159848.1 | - | 2 | 140 | |||
AK166670.1 | - | 2 | 143 | |||
AK168347.1 | - | 2 | 143 | |||
AK169101.1 | - | 2 | 143 | |||
AK171892.1 | - | 2 | 143 | |||
AK209547.1 | - | 1 | 16 | |||
AK220299.1 | - | 2 | 168 | |||
BC017539.1 | - | 2 | 138 | |||
BC035525.1 | - | 2 | 143 | |||
BC054785.1 | - | 1610 | ||||
BC065172.1 | - | 2 | 138 | 6415170 |