Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF376726.1 | - | 4 | 346 | ||
AK047364.1 | - | 4 | 322 | |||
AK082797.1 | - | 4 | 351 | |||
AK159313.1 | - | 4 | 370 | |||
AK170948.1 | - | 4 | 374 | |||
AK171030.1 | - | 4 | 374 | |||
AK184723.1 | - | 1 | 61 | |||
AK211611.1 | - | 1 | 61 | |||
AX775895.1 | - | 4 | 382 | |||
AX775897.1 | - | 4 | 346 | |||
BB611826.1 | - | 4 | 400 | |||
BC016104.1 | - | 4 | 351 | 4920840 | ||
BG079632.2 | - | 4 | 386 |