Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027456.1 | NP_081732.1 | 8 | 764 | ||
NM_030184.2 | NP_084460.1 | 8 | 799 | |||
GenBank mRNA | AK014450.1 | - | 8 | 768 | ||
AK017564.1 | - | 8 | 765 | |||
AK019600.1 | - | 8 | 788 | |||
AK030307.1 | - | 8 | 796 | |||
AK036083.1 | - | 8 | 770 | |||
AK039800.1 | - | 8 | 772 | |||
AK045699.1 | - | 8 | 763 | |||
AK046693.1 | - | 8 | 782 | |||
AK048064.1 | - | 8 | 778 | |||
AK140598.1 | - | 8 | 795 | |||
AK161393.1 | - | 8 | 779 | |||
AK164041.1 | - | 8 | 798 | |||
BC043101.1 | - | 8 | 751 | 6406321 |