Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030720.1 | NP_109645.1 | 2 | 1236 | ||
GenBank mRNA | AF272948.1 | - | 2 | 1236 | ||
BC023249.1 | - | 2 | 1245 | 3995193 |
Set Membership: |
![]() |