Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252580.1 | NP_001239509.1 | 11 | 1635 | ||
NM_019729.3 | NP_062703.2 | 11 | 1602 | |||
GenBank mRNA | AF057146.1 | - | 10 | 1393 | ||
AK122195.1 | - | 11 | 1513 | |||
AK129003.1 | - | 11 | 1603 | |||
AK134058.1 | - | 11 | 1557 | |||
AK142881.1 | - | 8 | 1100 | |||
AK153015.1 | - | 10 | 1350 | |||
AK163469.1 | - | 11 | 1535 | |||
BC027052.1 | - | 1 | 155 | |||
BC038123.1 | - | 11 | 1587 | |||
BC050947.1 | - | 11 | 1567 | 6825981 | ||
BC061465.1 | - | 11 | 1567 | 6825981 | ||
BC066126.1 | - | 11 | 1565 | 30301665 |