Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2l13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153516.2 | NP_705736.1 | 5 | 578 | ||
GenBank mRNA | AK031018.1 | - | 5 | 527 | ||
AK047223.1 | - | 5 | 580 | |||
AK088417.1 | - | 5 | 593 | |||
AK132774.1 | - | 5 | 587 | |||
AK143457.1 | - | 5 | 591 | |||
AK148007.1 | - | 5 | 597 | |||
AK165068.1 | - | 5 | 571 | |||
AK168833.1 | - | 5 | 568 | |||
AK178590.1 | - | 4 | 351 | |||
AK179659.1 | - | 4 | 350 | |||
AK186867.1 | - | 4 | 352 | |||
BC027307.1 | - | 5 | 468 | 4014844 | ||
BC029016.1 | - | 5 | 578 | 5358785 |