Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104.3 | NP_001095.2 | 15, 16 | 1955 | ||
NM_001258371.2 | NP_001245300.2 | 15, 16 | 2103 | |||
NR_047663.2 | 15, 16 | 1955 | ||||
GenBank mRNA | AK125851.1 | - | 15, 16 | 2489 | ||
AK303044.1 | - | 15, 16 | 2103 | |||
AK313556.1 | - | 15, 16 | 1893 | |||
BC096323.2 | - | 9, 10 | 1015 | |||
BC099647.3 | - | 15, 16 | 1876 | 40008220 | ||
BC099648.1 | - | 5, 6 | 624 | |||
BC099649.3 | - | 15, 16 | 1876 | 40008223 | ||
DA827389.1 | - | 4, 5 | 539 | |||
M86407.1 | - | 15, 16 | 1876 |