Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271347.1 | NP_001258276.1 | 4 | 724 | ||
NM_001271348.1 | NP_001258277.1 | 4 | 685 | |||
NM_001271349.1 | NP_001258278.1 | 3 | 622 | |||
NM_134015.3 | NP_598776.1 | 5 | 787 | |||
GenBank mRNA | AB093260.1 | - | 3 | 429 | ||
AK041532.1 | - | 4 | 517 | |||
AK149139.1 | - | 4 | 507 | |||
AK152181.1 | - | 4 | 447 | |||
AK160086.1 | - | 3 | 423 | |||
AK166226.1 | - | 4 | 382 | |||
AK168667.1 | - | 3 | 439 | |||
AK218380.1 | - | 2 | 112 | |||
AY038079.1 | - | 5 | 451 | |||
BC034261.1 | - | 5 | 467 | 4237375 |