Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291186.1 | NP_001278115.1 | 3 | 358 | ||
NM_198018.2 | NP_932135.1 | 7 | 1133 | |||
NM_198894.2 | NP_942597.1 | 8 | 1019 | |||
NM_198895.2 | NP_942598.1 | 8 | 1140 | |||
GenBank mRNA | AK052216.1 | - | 7 | 1351 | ||
AK134993.1 | - | 3 | 346 | |||
AK142364.1 | - | 8 | 1122 | |||
AK153615.1 | - | 7 | 1801 | |||
AK181053.1 | - | 1 | 54 | |||
AK191315.1 | - | 1 | 54 | |||
BC056385.1 | - | 3 | 347 | 6854368 | ||
BC058708.1 | - | 7 | 1133 | 6839300 | ||
BC059064.1 | - | 3 | 342 | 6825281 | ||
CB246381.1 | - | 3 | 358 |