Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other P2RY6 SIRNA Assays ›
          Gene Symbol:
          P2RY6
          Gene Name:
          pyrimidinergic receptor P2Y6
          Gene Aliases:
          P2Y6
          Chromosome Location:
          Chr.11: 73264503 - 73298625 on Build GRCh38
          UniGene ID:
          Hs.16362
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s9972
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: P2RY6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277204.1 NP_001264133.1 3 639
          NM_001277205.1 NP_001264134.1 4 586
          NM_001277206.1 NP_001264135.1 3 544
          NM_001277207.1 NP_001264136.1 3 486
          NM_001277208.1 NP_001264137.1 2 553
          NM_176796.2 NP_789766.1 4 619
          NM_176797.2 NP_789767.1 3 459
          NM_176798.2 NP_789768.1 4 736
          XM_005274022.3 XP_005274079.1 2 2727
          XM_006718571.3 XP_006718634.1 3 2812
          XM_011545076.2 XP_011543378.1 3 1942
          XM_011545077.2 XP_011543379.1 4 2061
          XM_011545079.2 XP_011543381.1 2 375
          GenBank mRNA AF007891.1 - 3 425
          AF007892.1 - 4 736
          AF007893.1 - 3 1259
          AF218005.1 - 3 496
          AF498920.1 - 1 194
          AL520218.3 - 4 607
          AL525099.3 - 4 581
          AX775903.1 - 4 736
          BC000571.2 - 2 434 3162734
          BC009391.2 - 2 493 4128941
          BQ718871.1 - 3 564
          BT006771.1 - 1 194
          BX337204.2 - 3 486
          BX381745.2 - 4 607
          BX397649.2 - 3 533
          U52464.1 - 3 397
          X97058.1 - 3 470

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Matching Taqman Gene Expression Assay:


          Hs00602548_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          phospholipase C-activating G-protein coupled receptor signaling pathway
          positive regulation of smooth muscle cell migration
          transepithelial chloride transport
          G-protein coupled purinergic nucleotide receptor signaling pathway
          cellular response to prostaglandin E stimulus
          cellular response to organic cyclic compound
          G-protein coupled receptor activity
          protein binding
          G-protein coupled purinergic nucleotide receptor activity
          UDP-activated nucleotide receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6f7b98477d-hxtzg:80/100.66.130.62:80.
          git-commit: 5163bf559e7ad0ebe9326a88094353fc6888890d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline