Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53017. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Catalyzes the initial reaction in O-linked oligosaccharide biosynthesis, the transfer of an N-acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GalNAc-T-like protein 4; GalNAc-T18; GalNAc-transferase 18; Polypeptide GalNAc transferase 18; Polypeptide GalNAc transferase-like protein 4; Polypeptide N-acetylgalactosaminyltransferase 18; Polypeptide N-acetylgalactosaminyltransferase-like protein 4; pp-GaNTase-like protein 4; Protein-UDP acetylgalactosaminyltransferase-like protein 4; putative polypeptide N-acetylgalactosaminyltransferase-like protein 4; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like protein 4; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 15; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 18; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase-like 4
Gene Aliases: GalNAc-T15; GalNAc-T18; GALNACT18; GALNT15; GALNT18; GALNTL4
UniProt ID: (Human) Q6P9A2
Entrez Gene ID: (Human) 374378
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support