Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57652. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
RBM27 gene ontology annotations related to this gene include mRNA processing; positive regulation of RNA export from nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acidic rich RS domain containing 1; RNA-binding motif protein 27; RNA-binding protein 27
Gene Aliases: ARRS1; KIAA1311; Psc1; RBM27; ZC3H18; ZC3H20
UniProt ID: (Human) Q9P2N5
Entrez Gene ID: (Human) 54439
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support