Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%).
This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
CSL plays a role in the regulatory network through which muscle cells coordinate their structural and functional states during growth, adaptation, and repair. The protein is abundantly expressed in heart and skeletal muscle and expression is increased by passive stretch. Mutations in this gene can lead to X-linked deafness.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CBF-1; H-2K binding factor-2; immunoglobulin kappa J region recombination signal binding protein 1; J kappa-recombination signal-binding protein; RBP-J; RBP-J kappa; RBP-JK; rbp-jkappa; Recombining binding protein suppressor of hairless; Renal carcinoma antigen NY-REN-30; suppressor of hairless homolog
Gene Aliases: AOS3; CBF1; csl; IGKJRB; IGKJRB1; KBF2; RBP-J; RBPJ; RBPJK; RBPSUH; SUH
UniProt ID: (Human) Q06330
Entrez Gene ID: (Human) 3516
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support