Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%).
This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
RUNX1T1 is a putative zinc finger transcription factor and oncoprotein. In acute myeloid leukemia, especially in the M2 subtype, the t(8;21)(q22;q22) translocation is one of the most frequent karyotypic abnormalities. The translocation produces a chimeric gene made up of the 5'-region of the RUNX1 gene fused to the 3'-region of this gene. The chimeric protein is thought to associate with the nuclear corepressor/histone deacetylase complex to block hematopoietic differentiation. The protein encoded by this gene is a putative zinc finger transcription factor and oncoprotein. In acute myeloid leukemia, especially in the M2 subtype, the t(8;21)(q22;q22) translocation is one of the most frequent karyotypic abnormalities. The translocation produces a chimeric gene made up of the 5'-region of the RUNX1 gene fused to the 3'-region of this gene. The chimeric protein is thought to associate with the nuclear corepressor/histone deacetylase complex to block hematopoietic differentiation. Several transcript variants encoding multiple isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acute myelogenous leukemia 1 translocation 1, cyclin-D related; CBFA2T1 isoform r1t1-11a61; CBFA2T1 isoform r1t1-11a62; CBFA2T1 isoform r1t1-11a63; CBFA2T1 isoform r1t1-11a64; CBFA2T1 isoform r1t1-11a65; CBFA2T1 isoform r1t1-7a47; CBFA2T1 isoform r1t1-7a48; CBFA2T1 isoform r1t1-7a49; CBFA2T1 isoform r1t1-7a50; CBFA2T1 isoform r1t1-7a51; CBFA2T1 isoform r1t1-7a52; CBFA2T1 isoform r1t1-7d53; CBFA2T1 isoform r1t1-7d54; CBFA2T1 isoform r1t1-7d55; CBFA2T1 isoform r1t1-7d56; CBFA2T1 isoform r1t1-8a57; CBFA2T1 isoform r1t1-8a58; CBFA2T1 isoform r1t1-8a59; CBFA2T1 isoform r1t1-8a60; core-binding factor, runt domain, alpha subunit 2; translocated to, 1; cyclin D-related; Cyclin-D-related protein; Eight twenty one protein; MGC2796; MTG8b; myeloid translocation gene on 8q22; Protein CBFA2T1; Protein ETO; Protein MTG8; runt related transcription factor 1; translocated to, 1 (cyclin D related); runt-related transcription factor 1; translocated to, 1 (cyclin D-related); Zinc finger MYND domain-containing protein 2
Gene Aliases: AML1-MTG8; AML1T1; CBFA2T1; CDR; ETO; MTG8; RUNX1T1; ZMYND2
UniProt ID: (Human) Q06455
Entrez Gene ID: (Human) 862
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support