Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67415. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
SH3BGRL3 could act as a modulator of glutaredoxin biological activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 297; SH3 domain binding glutamate-rich protein like 3; SH3 domain binding glutamic acid-rich protein like 3; SH3 domain-binding glutamic acid-rich-like protein 3; SH3 domain-binding protein 1; SH3BGRL3-like protein; SH3BP-1; TIP-B1; TNF inhibitory protein; TNF inhibitory protein B1
Gene Aliases: HEL-S-297; P1725; SH3BGRL3; SH3BP-1; TIP-B1
UniProt ID: (Human) Q9H299
Entrez Gene ID: (Human) 83442
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support