Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (68%), Rat (68%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83834. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
SPINT4 is a protein coding gene. Gene ontology (GO) annotation include extracellular region/cellular component.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kunitz-type protease inhibitor 4; serine protease inhibitor, Kunitz type 1; serine protease inhibitor, Kunitz type 4
Gene Aliases: C20orf137; dJ601O1.1; SPINT3; SPINT4
UniProt ID: (Human) Q6UDR6
Entrez Gene ID: (Human) 391253
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support