Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (47%), Rat (47%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67340. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
TGFBR3L is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TGF-beta receptor type III-like protein; TGF-beta receptor type-3-like protein; TGFR-3L; transforming growth factor beta receptor III like; transforming growth factor, beta receptor III-like; Transforming growth factor-beta receptor type 3-like protein; Transforming growth factor-beta receptor type III-like protein
Gene Aliases: TGFBR3L; TGFR-3L
UniProt ID: (Human) H3BV60
Entrez Gene ID: (Human) 100507588
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support