Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (65%), Rat (65%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64110. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Transmembrane protein 108 (TMEM108) is a gene associated with psychiatric disorders such as schizophrenia, bipolar disorder, and major depression disorder. It is specifically expressed in the nervous system, particularly enriched in the postsynaptic density fraction and DG granule neurons. Research has shown that TMEM108 is critical for adult neurogenesis in the hippocampal dentate gyrus, indicating its role in neuronal development and function. Studies with TMEM108 mutant mice exhibit mania-like behavior, highlighting its involvement in the pathology of bipolar disorder. The gene's expression is notably higher in oligodendrocyte lineage cells within the central nervous system compared to other lineage cells. As a schizophrenia susceptibility gene, TMEM108 has been identified as a potential target for understanding the underlying mechanisms of psychiatric diseases and for developing therapeutic approaches. However, the detailed pathophysiological mechanisms by which TMEM108 contributes to such disorders remain largely unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer/testis antigen 124; Retrolinkin; Transmembrane protein 108
Gene Aliases: CT124; KIAA1690; RTLN; TMEM108; UNQ1875/PRO4318
UniProt ID: (Human) Q6UXF1
Entrez Gene ID: (Human) 66000
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support