Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55863. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Core component of the DBIRD complex, a multiprotein complex that acts at the interface between core mRNP particles and RNA polymerase II (RNAPII) and integrates transcript elongation with the regulation of alternative splicing: the DBIRD complex affects local transcript elongation rates and alternative splicing of a large set of exons embedded in (A + T)-rich DNA regions. May play a role in neuronal differentiation and is able to bind DNA and activate expression in vitro. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DBIRD complex subunit ZNF326; dJ871E2.1 (novel protein (ortholog of mouse zinc finger protein Zan75)); Zinc finger protein 326; Zinc finger protein interacting with mRNPs and DBC1; ZNF-protein interacting with nuclear mRNPs and DBC1
Gene Aliases: dJ871E2.1; ZAN75; Zfp326; ZIRD; ZNF326
UniProt ID: (Human) Q5BKZ1
Entrez Gene ID: (Human) 284695
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support