Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (34%), Rat (34%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51625. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a protein that likely functions as a transcription factor. The protein contains an N-terminal KRAB domain and several C2H2-type zinc finger motifs. Mutations in this gene cause mental retardation X-linked type 45 (MRX45).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HFZ20; Zinc finger protein 81
Gene Aliases: dJ54B20.6; HFZ20; MRX45; ZNF81
UniProt ID: (Human) P51508
Entrez Gene ID: (Human) 347344
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support