The General Purpose Pro Centrifuges are a refrigerated 4L floor and bench models equipped with an intuitive touchscreen interface, Auto-Lock Rotor Exchange Technology and Biocontainment Lids.
General Purpose Pro Centrifuges are designed to meet the needs of today’s rapid-fire discoveries, with updates to help you get your work done more quickly, consistently, and with powerful reliability.
CTS Series Equipment Packages consist of the equipment, a documentation package and compliance services. These packages support your GMP needs for cell and gene therapy manufacturing. They help you get up and running faster, stay compliant, support regulatory audits, and stay on schedule.
CTS Series Equipment Package
A comprehensive product documentation package includes regulatory certificates, recommended protocols and product-specific factory documentation and specification.
Gain confidence with our compliance services that can help reduce your risk of non-compliant instruments. Service options include Installation Qualification (IQ), Operation Qualification (OQ) and cycle testing.
Choose your model and package
Models | Sorvall X4R Pro | Sorvall X4F R Pro | ||||||||
![]() | ![]() | |||||||||
|
| |||||||||
Model | • Benchtop | • Floor standing | ||||||||
Capacity | • 4L capacity | • 4L capacity | ||||||||
Rotor | → TX-1000 rotor | → TX-1000 rotor | ||||||||
Buckets | → 4 x 1000mL | → 4 x 1000mL | ||||||||
Lids | → Biocontainment Certified Lids | → Biocontainment Certified Lids | ||||||||
Adapters | → 50 ml conical adapters → 15 ml conical adapters → 500 mlconical adapters | → 50 ml conical adapters → 15 ml conical adapters → 500 mlconical adapters |
Compliance Services* (separate order number required) – Reduce equipment noncompliance by documenting functionality to support your regulatory requirements with qualification (IQ/OQ) and performance testing services.
Qualification service Cat. No.
On-site IQ/OQ service IOQPCKE89003492
On-site IQ/OQ + cycle test IOPQPCKE89003492
*Compliance services are only available in select countries including U.S., Germany, France, U.K., Ireland, Spain, Portugal, Italy, Austria, Switzerland, Norway, Sweden, Denmark, Netherlands. Please state your interest to the “Request a quote” form.
Models | Multifuge X4R Pro | Multifuge X4F R Pro | ||||||||
![]() | ![]() | |||||||||
|
| |||||||||
Model | • Benchtop | • Floor standing | ||||||||
Capacity | • 4L capacity | • 4L capacity | ||||||||
Rotor | → TX-1000 rotor | → TX-1000 rotor | ||||||||
Buckets | → 4 x 1000mL | → 4 x 1000mL | ||||||||
Lids | → Biocontainment Certified Lids | → Biocontainment Certified Lids | ||||||||
Adapters | → 50 ml conical adapters → 15 ml conical adapters → 500 mlconical adapters | → 50 ml conical adapters → 15 ml conical adapters → 500 mlconical adapters |
Compliance Services* (separate order number required) – Reduce equipment noncompliance by documenting functionality to support your regulatory requirements with qualification (IQ/OQ) and performance testing services.
Qualification service Cat. No.
On-site IQ/OQ service IOQPCKE89003492
On-site IQ/OQ + cycle test IOPQPCKE89003492
*Compliance services are only available in select countries including U.S., Germany, France, U.K., Ireland, Spain, Portugal, Italy, Austria, Switzerland, Norway, Sweden, Denmark, Netherlands. Please state your interest to the “Request a quote” form.
Models | Sorvall X4R Pro | Sorvall X4F R Pro | ||||||||
![]() | ![]() | |||||||||
|
| |||||||||
Model | • Benchtop | • Floor standing | ||||||||
Capacity | • 4L capacity | • 4L capacity | ||||||||
Rotor | → TX-1000 rotor | → TX-1000 rotor | ||||||||
Buckets | → 4 x 1000mL | → 4 x 1000mL | ||||||||
Lids | → Biocontainment Certified Lids | → Biocontainment Certified Lids | ||||||||
Adapters | → 50 ml conical adapters → 15 ml conical adapters → 500 mlconical adapters | → 50 ml conical adapters → 15 ml conical adapters → 500 mlconical adapters |
Compliance Services* (separate order number required) – Reduce equipment noncompliance by documenting functionality to support your regulatory requirements with qualification (IQ/OQ) and performance testing services.
Qualification service Cat. No.
On-site IQ/OQ service IOQPCKE89003492
On-site IQ/OQ + cycle test IOPQPCKE89003492
*Compliance services are only available in select countries including U.S., Germany, France, U.K., Ireland, Spain, Portugal, Italy, Austria, Switzerland, Norway, Sweden, Denmark, Netherlands. Please state your interest to the “Request a quote” form.
Models | Multifuge X4R Pro | Multifuge X4F R Pro | ||||||||
![]() | ![]() | |||||||||
|
| |||||||||
Model | • Benchtop | • Floor standing | ||||||||
Capacity | • 4L capacity | • 4L capacity | ||||||||
Rotor | → TX-1000 rotor | → TX-1000 rotor | ||||||||
Buckets | → 4 x 1000mL | → 4 x 1000mL | ||||||||
Lids | → Biocontainment Certified Lids | → Biocontainment Certified Lids | ||||||||
Adapters | → 50 ml conical adapters → 15 ml conical adapters → 500 mlconical adapters | → 50 ml conical adapters → 15 ml conical adapters → 500 mlconical adapters |
Compliance Services* (separate order number required) – Reduce equipment noncompliance by documenting functionality to support your regulatory requirements with qualification (IQ/OQ) and performance testing services.
Qualification service Cat. No.
On-site IQ/OQ service IOQPCKE89003492
On-site IQ/OQ + cycle test IOPQPCKE89003492
*Compliance services are only available in select countries including U.S., Germany, France, U.K., Ireland, Spain, Portugal, Italy, Austria, Switzerland, Norway, Sweden, Denmark, Netherlands. Please state your interest to the “Request a quote” form.
Enhanced features of general purpose pro centrifuges
User interface
Simple touchscreen programming
Discover the Sorvall X Pro touchscreen features including:
- Easily accessible menu bar
- Centrifuge health icon for at-a-glance monitoring
- Time-saving pre-temp button
- Quick spins
- Useful information icon for additional support
Rotor exchange
Auto-Lock Rotor Exchange
Helps save time for faster results:
- Secure, ergonomic push-button rotor exchange in as little as three seconds
- Trouble-free installation and removal with no tools required
- Full chamber access for cleaning convenience and minimizes risk of drip tray contamination
- Flexibility to quickly switch between applications and evolve with the changing needs of your laboratory
Carbon fiber rotors
Ergonomics with enhanced performance
• Fiberlite rotor/lid combinations offer protection to the user, centrifuge and samples during centrifugation*
• Carbon fiber construction supported by a 15 year Fiberlite Rotor warranty
• Fiberlite Rotors reach higher speeds than metal counterparts, improving separations and results
• Ergonomics driven by lightweight construction, integrated handle design and Auto-Lock Rotor exchange
*Biocontainment certified by National Institute for Health Protection, Porton Down, UK
Biocontainer lids
ClickSeal Biocontainment Lids
One-handed sample protection
- Biocontainment sealing options for glove-friendly, one-handed open/close capability are ergonomically designed for both right and left-handed operations
- Simple operation for all laboratory users, eliminating multi-turn screw caps and complicated high pressure clips
- Biocontainment certification by National Institute for Health Protection, Porton Down, UK
Safety and ergonomics
- Accessory tray holds tools or samples as you prepare for your run.
- Front USB port for easier plug in and data transfer
Enhancements
- Accessory tray holds tools or samples as you prepare for your run.
- Front USB port for easier plug in and data transfer