Thermo Scientific™
Niton™ XL5 Plus Handheld XRF Analyzer
Catalog number: NITONXL5PLUS
Confidently perform elemental analysis with the Thermo Scientific™ Niton™ XL5 Plus handheld XRF analyzer; it delivers unprecedented performance even for lightweight elements. Identify pure metals and alloys, detect tramp elements, or obtain geochemical data. Industrial businesses rely on the Niton XL5 Plus analyzer for more than just low detection limits. It generates fast results and offers great versatility such as the ability to scan a broad range of materials with a single analyzer.
Features
  • 5W X-ray tube
  • Proprietary large area drift detector for light element detection (Mg, Al, Si, P, S)
  • Lightweight design (2.8 lbs/ 1.3 kg)
  • Miniaturized nose
  • WiFi enabled
  • Macro and micro cameras
  • Hot swap battery
  • Tilting, color touchscreen
  • Directional keys
  • Password protected security
  • IP54 certified (splash/dust proof)
Thermo Scientific™
Niton™ XL3t GOLDD+ XRF Analyzer
Catalog number: XL3TGOLDDPLUS
Industries from mining and exploration to scrap metal recycling depend on fast, accurate elemental analysis. The Thermo Scientific™ Niton™ XL3t GOLDD+ XRF Analyzer provides reliable, real-time data for sample identification and positive material identification for even difficult-to-analyze light elements, without helium purging or vacuum pumps. The Niton XL3t GOLDD+ Analyzer delivers a feature-rich package enhanced with Geometrically Optimized Large Area Drift Detector (GOLDD™) technology for improved light element (Mg-S) analysis, the lowest limits of detection and fastest measurement times.
Performance
GOLDD Technology: When low detection limits and high sample throughput are critical, the Niton XL3t GOLDD+ XRF analyzer delivers measurement times up to 10-times faster than conventional Si-PIN detectors and up to 3-times more precise than conventional smaller silicon drift detectors (SDD) - without helium purge or vacuum. For further enhancing light element analysis, a helium purge option is available. Niton analyzers also feature the highest performance X-ray tube and speed detector available on the market.
Features
  • Standard analytical range: Up to 30 elements from Mg to U (varies by application).
  • Most powerful x-ray tube on the market for handheld XRF analysis.
  • Point and shoot simplicity- very easy to use even by nontechnical personnel.
  • Rugged design for real-world industrial environments.
  • Nondestructive analysis with near instantaneous results.
  • Sealed against moisture and dust.
  • Ergonomically designed.
  • Daylight-readable icons.
  • Incorporates multi-language options.
  • Color, touch-screen display