Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT5G14710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121475.4 | NP_196975.1 | 1 - 2 | 261 | 103 | |
GenBank mRNA | AK117635.1 | - | 1 - 2 | 163 | 103 | |
AY084903.1 | - | 1 - 2 | 137 | 103 | ||
BT005169.1 | - | 1 - 2 | 60 | 103 |
Set Membership: |
![]() ![]() ![]() ![]() |