Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CID3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104295.4 | NP_175819.1 | 13 - 14 | 1984 | 89 | |
GenBank mRNA | AY136398.1 | - | 13 - 14 | 1875 | 89 | |
BT000204.1 | - | 12 - 13 | 1667 | 89 |
Set Membership: |
![]() ![]() ![]() ![]() |