Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104315.5 | NP_175839.2 | 17 - 18 | 1270 | 77 | |
GenBank mRNA | AK226435.1 | - | 17 - 18 | 1170 | 77 |
Set Membership: |
![]() ![]() ![]() ![]() |