Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS04 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104793.3 | NP_564772.1 | 11 - 12 | 2112 | 113 | |
GenBank mRNA | AY059757.1 | - | 11 - 12 | 2110 | 113 | |
BT001960.1 | - | 11 - 12 | 2058 | 113 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |