Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124823.2 | NP_001118295.1 | 11 - 12 | 1613 | 112 | |
NM_001335364.1 | NP_001324918.1 | 11 - 12 | 1588 | 112 | ||
NM_001335365.1 | NP_001324917.1 | 11 - 12 | 1507 | 112 | ||
NM_126831.4 | NP_178872.2 | 11 - 12 | 1670 | 112 |
Set Membership: |
![]() ![]() ![]() ![]() |