Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRT1.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332743.1 | NP_001321840.1 | 2 - 3 | 1262 | 68 | |
NM_102470.3 | NP_174028.2 | 3 - 4 | 1191 | 68 | ||
GenBank mRNA | EU266070.1 | - | 3 - 4 | 853 | 68 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |