Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIRO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036876.2 | NP_001031953.1 | 2 - 3 | 436 | 65 | |
NM_122636.4 | NP_198106.1 | 2 - 3 | 395 | 65 | ||
GenBank mRNA | AY081280.1 | - | 2 - 3 | 387 | 65 | |
BT003352.1 | - | 1 - 2 | 223 | 65 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |