Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174525.2 | NP_776950.1 | 2 - 3 | 115 | 54 | |
GenBank mRNA | AY645777.1 | - | 2 - 3 | 78 | 54 | |
M60328.1 | - | 2 - 3 | 115 | 54 |
Set Membership: |
![]() ![]() ![]() ![]() |