Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Bt03216868_g1
          See other HMGB1 GE Assays ›
          Gene Symbol:
          HMGB1
          Gene Name:
          high mobility group box 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 30327844 - 30333009 on Build ARS-UCD1.2
          Species:
          Bovine
          Species Specific ID (Flybase ID):
          -
          Assay ID Bt03216868_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HMGB1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_176612.1 NP_788785.1 1 - 2 97 65
          GenBank mRNA BC102929.1 - 1 - 2 83 65
          BT030587.1 - 1 - 2 118 65
          X12796.1 - 97 65

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC102929.1, NM_176612.1, X12796.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          eye development
          myeloid dendritic cell activation
          endothelial cell proliferation
          adaptive immune response
          plasmacytoid dendritic cell activation
          macrophage activation involved in immune response
          B cell proliferation involved in immune response
          dendritic cell chemotaxis
          regulation of tolerance induction
          regulation of T cell mediated immune response to tumor cell
          DNA topological change
          base-excision repair
          DNA recombination
          nucleosome assembly
          chromatin remodeling
          regulation of transcription by RNA polymerase II
          autophagy
          inflammatory response
          negative regulation of RNA polymerase II transcription preinitiation complex assembly
          lung development
          activation of protein kinase activity
          DNA geometric change
          tumor necrosis factor production
          positive regulation of interferon-alpha production
          positive regulation of interferon-beta production
          V(D)J recombination
          positive regulation of toll-like receptor 2 signaling pathway
          positive regulation of toll-like receptor 4 signaling pathway
          toll-like receptor 9 signaling pathway
          positive regulation of toll-like receptor 9 signaling pathway
          endothelial cell chemotaxis
          apoptotic cell clearance
          innate immune response
          positive regulation of myeloid cell differentiation
          positive regulation of glycogen catabolic process
          positive chemotaxis
          DNA ligation involved in DNA repair
          positive regulation of DNA ligation
          response to glucocorticoid
          cellular response to lipopolysaccharide
          positive regulation of monocyte chemotactic protein-1 production
          positive regulation of wound healing
          neutrophil clearance
          cellular response to interleukin-7
          positive regulation of sprouting angiogenesis
          regulation of nucleotide-excision repair
          four-way junction DNA binding
          bubble DNA binding
          damaged DNA binding
          double-stranded DNA binding
          single-stranded DNA binding
          double-stranded RNA binding
          single-stranded RNA binding
          cytokine activity
          DNA binding, bending
          calcium-dependent protein kinase regulator activity
          protein kinase activator activity
          chemoattractant activity
          histone binding
          non-sequence-specific DNA binding, bending
          unmethylated CpG binding
          RAGE receptor binding
          supercoiled DNA binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0