Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Bt03217446_m1
          See other EDN1 GE Assays ›
          Gene Symbol:
          EDN1
          Gene Name:
          endothelin 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.23: 44156440 - 44163423 on Build ARS-UCD1.2
          Species:
          Bovine
          Species Specific ID (Flybase ID):
          -
          Assay ID Bt03217446_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: EDN1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181010.2 NP_851353.1 2 - 3 527 60
          GenBank mRNA BC102459.1 - 2 - 3 547 60
          S37093.1 - 2 - 3 527 60
          X52740.1 - 277 60
          X52942.1 - 389 60

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          prostaglandin biosynthetic process
          branching involved in blood vessel morphogenesis
          response to hypoxia
          in utero embryonic development
          regulation of systemic arterial blood pressure by endothelin
          regulation of pH
          cell surface receptor signaling pathway
          adenylate cyclase-inhibiting G protein-coupled receptor signaling pathway
          protein kinase C-activating G protein-coupled receptor signaling pathway
          heart development
          respiratory gaseous exchange by respiratory system
          body fluid secretion
          regulation of blood pressure
          dorsal/ventral pattern formation
          positive regulation of heart rate
          regulation of glucose transmembrane transport
          neural crest cell development
          phosphatidylinositol 3-kinase signaling
          artery smooth muscle contraction
          vein smooth muscle contraction
          regulation of vasoconstriction
          calcium-mediated signaling
          peptide hormone secretion
          nitric oxide transport
          positive regulation of cell migration
          phospholipase D-activating G protein-coupled receptor signaling pathway
          negative regulation of cellular protein metabolic process
          positive regulation of urine volume
          positive regulation of renal sodium excretion
          protein kinase C deactivation
          middle ear morphogenesis
          rhythmic excitation
          positive regulation of JUN kinase activity
          positive regulation of cell size
          positive regulation of mitotic nuclear division
          positive regulation of transcription by RNA polymerase II
          positive regulation of smooth muscle contraction
          positive regulation of hormone secretion
          inositol phosphate-mediated signaling
          positive regulation of smooth muscle cell proliferation
          positive regulation of DNA-binding transcription factor activity
          cartilage development
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G protein-coupled signaling pathway
          negative regulation of nitric-oxide synthase biosynthetic process
          positive regulation of sarcomere organization
          positive regulation of prostaglandin-endoperoxide synthase activity
          positive regulation of cell growth involved in cardiac muscle cell development
          endothelin receptor signaling pathway
          cytokine activity
          hormone activity
          endothelin A receptor binding
          endothelin B receptor binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0