Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035301.1 | NP_001030378.1 | 2 - 3 | 472 | 62 | |
GenBank mRNA | BC113220.1 | - | 2 - 3 | 420 | 62 | |
BT021182.1 | - | 2 - 3 | 472 | 62 | ||
BT026299.1 | - | 2 - 3 | 339 | 62 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |