Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097988.1 | NP_001091457.1 | 2 - 3 | 193 | 133 | |
GenBank mRNA | BC140583.1 | - | 2 - 3 | 193 | 133 | |
BT030685.1 | - | 2 - 3 | 299 | 133 |
Set Membership: |
![]() ![]() ![]() ![]() |