Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099094.2 | NP_001092564.1 | 5 - 6 | 808 | 107 | |
GenBank mRNA | BC142252.1 | - | 5 - 6 | 809 | 107 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |