Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174769.2 | NP_777194.1 | 149 | 113 | ||
GenBank mRNA | U89439.1 | - | 149 | 113 |
Set Membership: |
![]() ![]() ![]() |