Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102523.2 | NP_001095993.1 | 2 | 1118 | 88 | |
GenBank mRNA | BC149904.1 | - | 2 | 1120 | 88 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |