Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Bt04945944_m1
          See other RACK1 GE Assays ›
          Gene Symbol:
          RACK1
          Gene Name:
          receptor for activated C kinase 1
          Gene Aliases:
          GNB2L1
          Chromosome Location:
          Chr.7: 40371008 - 40375831 on Build ARS-UCD1.2
          Species:
          Bovine
          Species Specific ID (Flybase ID):
          -
          Assay ID Bt04945944_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RACK1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175802.3 NP_786996.1 6 - 7 903 109
          GenBank mRNA BC102286.1 - 6 - 7 884 109
          EE233795.1 - 2 - 3 269 109
          EV698190.1 - 2 - 3 251 109
          EW687929.1 - 2 - 3 251 109

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cell cycle
          gastrulation
          negative regulation of gene expression
          protein ubiquitination
          negative regulation of Wnt signaling pathway
          negative regulation of cell growth
          positive regulation of cell migration
          positive regulation of protein-containing complex assembly
          negative regulation of protein binding
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          regulation of protein localization
          negative regulation of peptidyl-serine phosphorylation
          positive regulation of Golgi to plasma membrane protein transport
          positive regulation of apoptotic process
          pigmentation
          positive regulation of GTPase activity
          rhythmic process
          negative regulation of phagocytosis
          regulation of cell division
          positive regulation of cyclic-nucleotide phosphodiesterase activity
          regulation of cell cycle
          negative regulation of protein kinase B signaling
          positive regulation of mitochondrial depolarization
          negative regulation of protein tyrosine kinase activity
          cellular response to glucose stimulus
          cellular response to growth factor stimulus
          rescue of stalled ribosome
          negative regulation of hydrogen peroxide-induced neuron death
          regulation of establishment of cell polarity
          positive regulation of gastrulation
          positive regulation of intrinsic apoptotic signaling pathway
          protein kinase C binding
          ion channel inhibitor activity
          cysteine-type endopeptidase activator activity involved in apoptotic process
          protein phosphatase binding
          protein tyrosine kinase inhibitor activity
          cyclin binding
          receptor tyrosine kinase binding
          signaling adaptor activity
          SH2 domain binding
          protein homodimerization activity
          ribosome binding
          BH3 domain binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0