Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhx-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063213.7 | NP_495614.4 | 9 - 10 | 1646 | 155 | |
GenBank mRNA | AF497824.1 | - | 8 - 9 | 1626 | 155 |
Set Membership: |
![]() ![]() ![]() ![]() |