Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267957.3 | NP_001254886.1 | 24 - 25 | 6091 | 68 | |
NM_001381816.2 | NP_001366699.1 | 23 - 24 | 6025 | 68 |
Set Membership: |
![]() ![]() ![]() ![]() |