Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Ce02469393_m1
          See other EGL-8 GE Assays ›
          Gene Symbol:
          egl-8
          Gene Name:
          protein-coding
          Gene Aliases:
          CELE_B0348.4
          Chromosome Location:
          Chr.V: 20818 - 55451 on Build WBcel235
          Species:
          Caenorhabditis elegans
          Species Specific ID (Flybase ID):
          -
          Assay ID Ce02469393_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: egl-8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001026175.4 NP_001021346.1 14 - 15 3551 85
          NM_001026176.3 NP_001021347.1 14 - 15 3551 85
          NM_001313085.3 NP_001300014.1 15 3612 85
          NM_001313087.3 NP_001300016.1 14 - 15 3476 85
          NM_001313089.4 NP_001300018.1 13 - 14 3240 85
          NM_001313091.3 NP_001300020.1 13 - 14 3302 85
          NM_001313093.3 NP_001300022.1 12 - 13 3059 85
          NM_001313095.4 NP_001300024.1 14 3663 85
          NM_001313097.3 NP_001300026.1 13 3285 85
          NM_001313099.3 NP_001300028.1 13 3210 85
          NM_001313101.3 NP_001300030.1 12 2967 85
          NM_001313103.4 NP_001300032.1 12 - 13 3212 85
          NM_001313105.3 NP_001300034.1 11 2787 85
          NM_001372980.2 NP_001360389.1 13 - 14 3239 85
          NM_001372988.2 NP_001360397.1 11 - 12 2975 85
          NM_001380576.2 NP_001368552.1 15 - 16 3612 85
          NM_001380577.1 NP_001368553.1 14 - 15 3482 85
          NM_001380578.2 NP_001368554.1 13 - 14 3300 85
          NM_001380579.1 NP_001368555.1 12 - 13 3059 85
          NM_001380580.2 NP_001368556.1 14 - 15 3712 85
          NM_001380581.2 NP_001368557.1 13 - 14 3473 85
          NM_001380582.2 NP_001368558.1 13 - 14 3380 85
          NM_001380583.1 NP_001368559.1 12 - 13 3130 85
          NM_001380584.1 NP_001368560.1 12 - 13 3030 85
          GenBank mRNA AF179426.1 - 14 - 15 3551 85

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          chemotaxis
          signal transduction
          dopamine receptor signaling pathway
          positive regulation of acetylcholine secretion, neurotransmission
          lipid catabolic process
          protein localization to synapse
          intracellular signal transduction
          regulation of locomotion
          positive regulation of locomotion
          regulation of pharyngeal pumping
          thermotaxis
          regulation of oviposition
          phosphatidylinositol-mediated signaling
          positive regulation of axon regeneration
          phosphatidylinositol phospholipase C activity
          calcium ion binding
          phosphoric diester hydrolase activity
          hydrolase activity
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0