Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ce02469398_g1
See other EGL-8 GE Assays ›
Gene Symbol:
egl-8
Gene Name:
protein-coding
Gene Aliases:
CELE_B0348.4
Chromosome Location:
Chr.V: 20818 - 55451 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID
Ce02469398_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: egl-8
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001026175.4
NP_001021346.1
18 - 19
4189
88
NM_001026176.3
NP_001021347.1
19 - 20
4225
88
NM_001313085.3
NP_001300014.1
19
4286
88
NM_001313087.3
NP_001300016.1
19 - 20
4150
88
NM_001313089.4
NP_001300018.1
18 - 19
3914
88
NM_001313091.3
NP_001300020.1
18 - 19
3976
88
NM_001313093.3
NP_001300022.1
17 - 18
3733
88
NM_001313095.4
NP_001300024.1
19
4337
88
NM_001313097.3
NP_001300026.1
17
3959
88
NM_001313099.3
NP_001300028.1
17
3884
88
NM_001313101.3
NP_001300030.1
16
3641
88
NM_001313103.4
NP_001300032.1
17 - 18
3886
88
NM_001313105.3
NP_001300034.1
15
3461
88
NM_001372980.2
NP_001360389.1
17 - 18
3877
88
NM_001372988.2
NP_001360397.1
15 - 16
3613
88
NM_001380576.2
NP_001368552.1
19 - 20
4250
88
NM_001380577.1
NP_001368553.1
18 - 19
4120
88
NM_001380578.2
NP_001368554.1
17 - 18
3938
88
NM_001380579.1
NP_001368555.1
16 - 17
3697
88
NM_001380580.2
NP_001368556.1
18 - 19
4350
88
NM_001380581.2
NP_001368557.1
17 - 18
4111
88
NM_001380582.2
NP_001368558.1
17 - 18
4018
88
NM_001380583.1
NP_001368559.1
16 - 17
3768
88
NM_001380584.1
NP_001368560.1
16 - 17
3668
88
GenBank mRNA
AF179426.1
-
19 - 20
4225
88
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
Detect Genomic DNA
Caenorhabditis elegans
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
chemotaxis
signal transduction
dopamine receptor signaling pathway
positive regulation of acetylcholine secretion, neurotransmission
lipid catabolic process
protein localization to synapse
intracellular signal transduction
regulation of locomotion
positive regulation of locomotion
regulation of pharyngeal pumping
thermotaxis
regulation of oviposition
phosphatidylinositol-mediated signaling
positive regulation of axon regeneration
phosphatidylinositol phospholipase C activity
calcium ion binding
phosphoric diester hydrolase activity
hydrolase activity
metal ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0