Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: K07C5.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073258.6 | NP_505659.1 | 7 - 8 | 1417 | 117 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |