Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Cf02650448_m1
          See other CC2D2B GE Assays ›
          Gene Symbol:
          CC2D2B
          Gene Name:
          coiled-coil and C2 domain containing 2B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.28: 9371454 - 9480331 on Build CanFam3.1
          Species:
          Dog
          Species Specific ID (Flybase ID):
          -
          Assay ID Cf02650448_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CC2D2B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_014108844.2 XP_013964319.1 24 - 25 2971 99
          XM_014108856.2 XP_013964331.1 16 - 17 1945 99
          XM_022411979.1 XP_022267687.1 29 - 30 3562 99
          XM_022411980.1 XP_022267688.1 29 - 30 3563 99
          XM_022411981.1 XP_022267689.1 29 - 30 3529 99
          XM_022411982.1 XP_022267690.1 30 - 31 3617 99
          XM_022411983.1 XP_022267691.1 28 - 29 3463 99
          XM_022411985.1 XP_022267693.1 28 - 29 3448 99
          XM_022411986.1 XP_022267694.1 28 - 29 3433 99
          XM_022411987.1 XP_022267695.1 27 - 28 3420 99
          XM_022411988.1 XP_022267696.1 28 - 29 3364 99
          XM_022411989.1 XP_022267697.1 28 - 29 3352 99
          XM_022411990.1 XP_022267698.1 27 - 28 3334 99
          XM_022411991.1 XP_022267699.1 28 - 29 3441 99
          XM_022411992.1 XP_022267700.1 28 - 29 3418 99
          XM_022411993.1 XP_022267701.1 27 - 28 3377 99
          XM_022411994.1 XP_022267702.1 27 - 28 3250 99
          XM_022411997.1 XP_022267705.1 24 - 25 3114 99
          XM_022411998.1 XP_022267706.1 28 - 29 3473 99
          XM_022411999.1 XP_022267707.1 25 - 26 2887 99
          XM_022412000.1 XP_022267708.1 22 - 23 2889 99
          XM_022412001.1 XP_022267709.1 23 - 24 2800 99
          XM_022412002.1 XP_022267710.1 22 - 23 2746 99
          XM_022412003.1 XP_022267711.1 21 - 22 2679 99
          XM_022412006.1 XP_022267714.1 23 - 24 2664 99
          XM_022412007.1 XP_022267715.1 23 - 24 2706 99
          XM_022412008.1 XP_022267716.1 24 - 25 2751 99
          XM_022412009.1 XP_022267717.1 24 - 25 2736 99
          XM_022412010.1 XP_022267718.1 23 - 24 2690 99
          XM_022412011.1 XP_022267719.1 23 - 24 2707 99
          XM_022412012.1 XP_022267720.1 23 - 24 2668 99
          XM_022412013.1 XP_022267721.1 23 - 24 2651 99
          XM_022412014.1 XP_022267722.1 13 - 14 1583 99
          XM_022412015.1 XP_022267723.1 14 - 15 1754 99

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0