Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM200B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_849712.4 | XP_854805.1 | 2 | 721 | 88 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |