Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XKR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003638918.4 | XP_003638966.1 | 3 | 846 | 146 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |