Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dr03073246_g1
See other ELNA GE Assays ›
Gene Symbol:
elna
Gene Name:
elastin a
Gene Aliases:
eln1
Chromosome Location:
Chr.15: 263189 - 323096 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID
Dr03073246_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: elna
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001080063.2
NP_001073532.2
32 - 33
2220
75
XM_009291337.3
XP_009289612.1
32 - 33
2486
75
XM_017351021.2
XP_017206510.1
32 - 33
2487
75
XM_017351022.2
XP_017206511.1
32 - 33
2482
75
XM_017351023.2
XP_017206512.1
32 - 33
2482
75
XM_017351024.2
XP_017206513.1
32 - 33
2482
75
XM_017351025.2
XP_017206514.1
32 - 33
2487
75
XM_017351026.2
XP_017206515.1
32 - 33
2472
75
XM_017351027.2
XP_017206516.1
32 - 33
2487
75
XM_017351028.2
XP_017206517.1
32 - 33
2486
75
XM_017351029.2
XP_017206518.1
31 - 32
2442
75
XM_017351031.2
XP_017206520.1
30 - 31
2359
75
XM_017351032.2
XP_017206521.1
30 - 31
2338
75
XM_017351033.2
XP_017206522.1
28 - 29
2208
75
XM_017351035.2
XP_017206524.1
28 - 29
2209
75
XM_017351036.2
XP_017206525.1
28 - 29
2209
75
XM_017351037.2
XP_017206526.1
28 - 29
2209
75
XM_017351038.2
XP_017206527.1
24 - 25
1899
75
XM_021481472.1
XP_021337147.1
29 - 30
2445
75
XM_021481473.1
XP_021337148.1
26 - 27
2070
75
XM_021481474.1
XP_021337149.1
21 - 22
1857
75
GenBank mRNA
DQ523562.1
-
32 - 33
2220
75
MF039671.1
-
9 - 10
684
75
MF039672.1
-
9 - 10
684
75
MF039673.1
-
9 - 10
684
75
MF039674.1
-
9 - 10
684
75
MF039675.1
-
576
75
MF039677.1
-
3 - 4
186
75
MF101833.1
-
3 - 4
186
75
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Detect Genomic DNA
Danio rerio
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
extracellular matrix structural constituent
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0