Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03073258_g1
          See other ELNA GE Assays ›
          Gene Symbol:
          elna
          Gene Name:
          elastin a
          Gene Aliases:
          eln1
          Chromosome Location:
          Chr.15: 263189 - 323096 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03073258_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: elna
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001080063.2 NP_001073532.2 43 - 44 2815 93
          XM_009291337.3 XP_009289612.1 41 - 42 2988 93
          XM_017351021.2 XP_017206510.1 43 - 44 3178 93
          XM_017351022.2 XP_017206511.1 43 - 44 3173 93
          XM_017351023.2 XP_017206512.1 43 - 44 3173 93
          XM_017351024.2 XP_017206513.1 43 - 44 3173 93
          XM_017351025.2 XP_017206514.1 43 - 44 3166 93
          XM_017351026.2 XP_017206515.1 43 - 44 3163 93
          XM_017351027.2 XP_017206516.1 43 - 44 3154 93
          XM_017351028.2 XP_017206517.1 43 - 44 3177 93
          XM_017351029.2 XP_017206518.1 42 - 43 3133 93
          XM_017351031.2 XP_017206520.1 41 - 42 3050 93
          XM_017351032.2 XP_017206521.1 41 - 42 3029 93
          XM_017351033.2 XP_017206522.1 39 - 40 2899 93
          XM_017351035.2 XP_017206524.1 39 - 40 2900 93
          XM_017351036.2 XP_017206525.1 39 - 40 2900 93
          XM_017351037.2 XP_017206526.1 39 - 40 2900 93
          XM_017351038.2 XP_017206527.1 35 - 36 2590 93
          XM_021481472.1 XP_021337147.1 40 - 41 3136 93
          XM_021481473.1 XP_021337148.1 37 - 38 2761 93
          XM_021481474.1 XP_021337149.1 32 - 33 2548 93
          GenBank mRNA DQ523562.1 - 43 - 44 2815 93
          MF039657.1 - 8 - 9 403 93
          MF039659.1 - 6 - 7 310 93
          MF039660.1 - 8 - 9 499 93
          MF039661.1 - 8 - 9 403 93
          MF039662.1 - 8 - 9 403 93
          MF039663.1 - 8 - 9 403 93
          MF039666.1 - 3 - 4 180 93
          MF039667.1 - 3 - 4 276 93
          MF039668.1 - 1 - 2 87 93
          MF039670.1 - 3 - 4 180 93
          MF067533.1 - 3 - 4 143 93
          MF067534.1 - 3 - 4 155 93
          MF067535.1 - 3 - 4 148 93
          MF067537.1 - 3 - 4 143 93
          MF067538.1 - 3 - 4 143 93

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          extracellular matrix structural constituent

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0