Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03073269_m1
          See other ELNA GE Assays ›
          Gene Symbol:
          elna
          Gene Name:
          elastin a
          Gene Aliases:
          eln1
          Chromosome Location:
          Chr.15: 263189 - 323096 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03073269_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: elna
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001080063.2 NP_001073532.2 53 - 54 3354 135
          XM_009291337.3 XP_009289612.1 52 - 53 3569 135
          XM_017351021.2 XP_017206510.1 54 - 55 3759 135
          XM_017351022.2 XP_017206511.1 54 - 55 3754 135
          XM_017351023.2 XP_017206512.1 54 - 55 3754 135
          XM_017351024.2 XP_017206513.1 54 - 55 3754 135
          XM_017351025.2 XP_017206514.1 54 - 55 3747 135
          XM_017351026.2 XP_017206515.1 54 - 55 3744 135
          XM_017351027.2 XP_017206516.1 54 - 55 3735 135
          XM_017351028.2 XP_017206517.1 53 - 54 3716 135
          XM_017351029.2 XP_017206518.1 53 - 54 3714 135
          XM_017351031.2 XP_017206520.1 52 - 53 3631 135
          XM_017351032.2 XP_017206521.1 52 - 53 3610 135
          XM_017351033.2 XP_017206522.1 50 - 51 3480 135
          XM_017351035.2 XP_017206524.1 50 - 51 3481 135
          XM_017351036.2 XP_017206525.1 50 - 51 3481 135
          XM_017351037.2 XP_017206526.1 50 - 51 3481 135
          XM_017351038.2 XP_017206527.1 46 - 47 3171 135
          XM_021481472.1 XP_021337147.1 51 - 52 3717 135
          XM_021481473.1 XP_021337148.1 48 - 49 3342 135
          XM_021481474.1 XP_021337149.1 43 - 44 3129 135
          GenBank mRNA DQ523562.1 - 53 - 54 3354 135
          MF067532.1 - 12 - 13 663 135
          MF067533.1 - 13 - 14 682 135
          MF067534.1 - 13 - 14 694 135
          MF067535.1 - 13 - 14 687 135
          MF067536.1 - 13 - 14 682 135
          MF067537.1 - 13 - 14 682 135
          MF067538.1 - 14 - 15 724 135

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Danio rerio Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          extracellular matrix structural constituent

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0