Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03074235_m1
          See other PTK2AB GE Assays ›
          Gene Symbol:
          ptk2ab
          Gene Name:
          protein tyrosine kinase 2ab
          Gene Aliases:
          fak, fak1a, ptk2, ptk2.1, ptk2a, wu:fa18c04, zgc:66114
          Chromosome Location:
          Chr.16: 30737278 - 30869776 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03074235_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ptk2ab
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_131796.1 NP_571871.1 25 - 26 2596 82
          XM_005158162.4 XP_005158219.1 26 - 27 3011 82
          XM_005158163.4 XP_005158220.1 25 - 26 2990 82
          XM_005158164.4 XP_005158221.1 24 - 25 2972 82
          XM_005158165.4 XP_005158222.1 24 - 25 2972 82
          XM_009292411.3 XP_009290686.1 26 - 27 3011 82
          XM_021466598.1 XP_021322273.1 27 - 28 3127 82
          XM_021466599.1 XP_021322274.1 27 - 28 2669 82
          XM_021466600.1 XP_021322275.1 27 - 28 3118 82
          XM_021466601.1 XP_021322276.1 27 - 28 3126 82
          XM_021466602.1 XP_021322277.1 28 - 29 2691 82
          XM_021466603.1 XP_021322278.1 26 - 27 3109 82
          XM_021466604.1 XP_021322279.1 26 - 27 3099 82
          XM_021466605.1 XP_021322280.1 26 - 27 3096 82
          XM_021466606.1 XP_021322281.1 25 - 26 3078 82
          XM_021466607.1 XP_021322282.1 26 - 27 3021 82
          XM_021466608.1 XP_021322283.1 26 - 27 3020 82
          XM_021466609.1 XP_021322284.1 28 - 29 2786 82
          GenBank mRNA AF348085.1 - 25 - 26 2596 82
          BC163886.1 - 23 - 24 2409 82

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          protein phosphorylation
          transmembrane receptor protein tyrosine kinase signaling pathway
          signal complex assembly
          epidermal growth factor receptor signaling pathway
          phosphorylation
          midbrain-hindbrain boundary morphogenesis
          cell differentiation
          regulation of cell adhesion
          peptidyl-tyrosine autophosphorylation
          regulation of cell population proliferation
          innate immune response
          lens fiber cell morphogenesis
          protein kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          signaling receptor binding
          ATP binding
          kinase activity
          transferase activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0